Merck
  • Histone deacetylase inhibitor SAHA-induced epithelial-mesenchymal transition by upregulating Slug in lung cancer cells.
Histone deacetylase inhibitor SAHA-induced epithelial-mesenchymal transition by upregulating Slug in lung cancer cells.
Anti-cancer drugs (2017-11-28)
Jun Wang, Mei-Qing Xu, Xian-Liang Jiang, Xin-Yu Mei, Xue-Gang Liu
ABSTRACT
SAHA, a member of histone deacetylase inhibitors (HDACIs), which emerged as a class of novel antitumor drug, has been used in clinical treatment of cancers. However, clinical experience of SAHA in solid tumors has been disappointing. Nevertheless, the underlying mechanism of this deficiency is not clearly understood. In the present study, we found that SAHA could induce epithelial-mesenchymal transitions (EMT) in lung cancer A549 cells, which was associated with increased migration capability and cellular morphology changes. We showed that SAHA decreased epithelial marker E-cadherin's expression and increased the expression of mesenchymal marker vimentin. SAHA upregulated the protein and mRNA expression of transcription factor Slug in a time-dependent manner and promoted its nuclear translocation. We further demonstrated that SAHA upregulated Slug expression by promoting Slug acetylation but not influencing the phosphorylation of GSK-3β, a main kinase-controlled Slug expression. Finally, silencing of Slug by siRNA reversed EMT marker expressions and cellular morphology change induced by SAHA, suggesting that Slug plays a crucial role in SAHA-mediated EMT in A549 cells. Our research study provided a better understanding of treatment failure of SAHA in patients with solid tumors. Therefore, more attention should be paid to cancer treatment using SAHA and strategies for reversing EMT before using SAHA would be better if the value of SAHA in the treatment of solid tumors, especially lung cancer, is realized.
Sigma-Aldrich
SAHA, ≥98% (HPLC)