Merck
  • Glutathione Enhances Antibiotic Efficiency and Effectiveness of DNase I in Disrupting
Glutathione Enhances Antibiotic Efficiency and Effectiveness of DNase I in Disrupting
Frontiers in microbiology (2018-01-10)
Theerthankar Das, Martin Simone, Amaye I Ibugo, Paul K Witting, Mike Manefield, Jim Manos
ABSTRACT
Pyocyanin secreted by