Merck
  • Mixed-Organic-Cation Tin Iodide for Lead-Free Perovskite Solar Cells with an Efficiency of 8.12.
Mixed-Organic-Cation Tin Iodide for Lead-Free Perovskite Solar Cells with an Efficiency of 8.12.
Advanced science (Weinheim, Baden-Wurttemberg, Germany) (2017-12-05)
Ziran Zhao, Feidan Gu, Yunlong Li, Weihai Sun, Senyun Ye, Haixia Rao, Zhiwei Liu, Zuqiang Bian, Chunhui Huang
ABSTRACT
In this work, a fully tin-based, mixed-organic-cation perovskite absorber (FA)