- Syntheses and characterization of phosphonates and diphosphonates of molybdenum, A
Syntheses and characterization of phosphonates and diphosphonates of molybdenum, A
Dalton transactions (Cambridge, England : 2003) (2017-11-11)
D Elias Jesu Packiam, Kanamaluru Vidyasagar
PMID29125165
ABSTRACT
Twenty new molybdenum phosphonates and diphosphonates have been synthesized and structurally characterized by single crystal and powder X-ray diffraction, CHN analyses, spectroscopic and thermal studies. Four of them are molecular phenyl- and benzyl-phosphonates containing discrete [(MoO