Merck
  • Biocompatibility and Osseointegration of Titanium Implants with a Silver-Doped Polysiloxane Coating: An In Vivo Pig Model.
Biocompatibility and Osseointegration of Titanium Implants with a Silver-Doped Polysiloxane Coating: An In Vivo Pig Model.
The International journal of oral & maxillofacial implants (2017-10-14)
Ralf Smeets, Clarissa Precht, Michael Hahn, Ole Jung, Philip Hartjen, Max Heiland, Alexander Gröbe, Marzellus Große Holthaus, Henning Hanken
ABSTRACT
To test the antimicrobial properties, surface topography, reaction of surrounding tissue (biocompatibility), and osseointegration of ultrathin implant surfaces containing polysiloxane and nanoscaled silver particles. Implants with polysiloxane coating and nanoscaled silver particles (Ag/SiO Roughness parameters did not differ between all three implant types. The Ag/SiO The osseointegration capability of the Ag/SiO