Merck
  • Effects of combination therapy using antithrombin and thrombomodulin for sepsis-associated disseminated intravascular coagulation.
Effects of combination therapy using antithrombin and thrombomodulin for sepsis-associated disseminated intravascular coagulation.
Annals of intensive care (2017-11-04)
Toshiaki Iba, Akiyoshi Hagiwara, Daizoh Saitoh, Hideaki Anan, Yutaka Ueki, Koichi Sato, Satoshi Gando
ABSTRACT
No single anticoagulant has been proven effective for sepsis-associated disseminated intravascular coagulation (DIC). Thus, the concomitant use of antithrombin concentrate and recombinant thrombomodulin has been conceived. This observational study was conducted to investigate the efficacy and safety of this combination therapy. A total of 510 septic DIC patients who received antithrombin substitution were retrospectively analyzed. Among them, 228 were treated with antithrombin and recombinant thrombomodulin (combination therapy) and the rest were treated with antithrombin alone (monotherapy). Propensity score matching created 129 matched pairs, and 28-day all-cause mortality, DIC scores, the sequential organ failure assessment (SOFA) scores, and the incidence of bleeding were compared. A log-rank test revealed a significant association between combination therapy and a lower 28-day mortality rate (hazard ratio 0.49, 95% confidence interval 0.29-0.82, P = 0.006) in the matched pairs. The DIC scores and the SOFA scores in the combination therapy group were significantly lower than those in the monotherapy group on Day 4 and Day 7. The incidence of bleeding did not differ between the groups (2.11 vs. 2.31%, P = 1.000). The current study demonstrated the potential benefit of adding recombinant thrombomodulin to antithrombin. The co-administration of these two anticoagulants was associated with reduced mortality among patients with sepsis-induced DIC without increasing the risk of bleeding.
Sigma-Aldrich
Thrombomodulin human, recombinant, expressed in HEK 293 cells, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture