Merck
  • Adenylyl Cyclase 1 Is Required for Ethanol-Induced Locomotor Sensitization and Associated Increases in NMDA Receptor Phosphorylation and Function in the Dorsal Medial Striatum.
Adenylyl Cyclase 1 Is Required for Ethanol-Induced Locomotor Sensitization and Associated Increases in NMDA Receptor Phosphorylation and Function in the Dorsal Medial Striatum.
The Journal of pharmacology and experimental therapeutics (2017-08-26)
Kelly E Bosse, Max F Oginsky, Laura L Susick, Sailesh Ramalingam, Carrie R Ferrario, Alana C Conti
ABSTRACT
Neuroadaptive responses to chronic ethanol, such as behavioral sensitization, are associated with
Sigma-Aldrich
NB001, ≥98% (HPLC)