Merck
  • Markers of neuroprotection of combined EPA and DHA provided by fish oil are higher than those of EPA (
Markers of neuroprotection of combined EPA and DHA provided by fish oil are higher than those of EPA (
Nutrition & metabolism (2017-10-14)
Paula A Lopes, Narcisa M Bandarra, Susana V Martins, Joana Martinho, Cristina M Alfaia, Marta S Madeira, Carlos Cardoso, Cláudia Afonso, Maria C Paulo, Rui M A Pinto, José L Guil-Guerrero, José A M Prates
ABSTRACT
To overcome the current overexploitation of fish rich in The experiment lasted for 10 weeks and involved 32 Wistar rats, divided into 4 diets ( Plasma lipid profile and dopamine levels were more beneficial in Fish Oil diet. In addition, Taken together, the combination of EPA and DHA from fish oil can counteract the undesirable health effects of saturated fat based diets and benefit, in the long run, neurological function.
Product Description
Supelco
Heneicosanoic acid, analytical standard
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)