Merck
  • Rhodoferax koreense sp. nov, an obligately aerobic bacterium within the family Comamonadaceae, and emended description of the genus Rhodoferax.
Rhodoferax koreense sp. nov, an obligately aerobic bacterium within the family Comamonadaceae, and emended description of the genus Rhodoferax.
Journal of microbiology (Seoul, Korea) (2017-09-29)
Mohamed El-Agamy Farh, Yeon-Ju Kim, Priyanka Singh, Sun Young Jung, Jong-Pyo Kang, Deok-Chun Yang
ABSTRACT
Gram-staining-negative, uniflagellated, rod-shaped, designated as DCY110
Sigma-Aldrich
Deoxyribonucleic acid sodium salt from Escherichia coli strain B, Genomic, unsheared