Merck
  • Oxidation Kinetics of Bromophenols by Nonradical Activation of Peroxydisulfate in the Presence of Carbon Nanotube and Formation of Brominated Polymeric Products.
Oxidation Kinetics of Bromophenols by Nonradical Activation of Peroxydisulfate in the Presence of Carbon Nanotube and Formation of Brominated Polymeric Products.
Environmental science & technology (2017-08-15)
Chaoting Guan, Jin Jiang, Suyan Pang, Congwei Luo, Jun Ma, Yang Zhou, Yi Yang
ABSTRACT
This work demonstrated that bromophenols (BrPs) could be readily oxidized by peroxydisulfate (PDS) activated by a commercial carbon nanotube (CNT), while furfuryl alcohol (a chemical probe for singlet oxygen (
Sigma-Aldrich
2,2,6,6-Tetramethyl-4-piperidinol, 98%
Sigma-Aldrich
4-Methoxyphenol, ReagentPlus®, 99%