Merck
  • Structure-activity profiles of macrolactam immunosuppressant FK-506 analogues.
Structure-activity profiles of macrolactam immunosuppressant FK-506 analogues.
FEBS letters (1993-01-25)
M Kawai, B C Lane, G C Hsieh, K W Mollison, G W Carter, J R Luly
ABSTRACT
The immunosuppressive agent FK-506 has received much attention due to its efficacy and potency in the areas of transplant rejection and autoimmune disease. Calcineurin, a Ca(2+)-calmodulin activated phosphatase, was recently implicated in the immunosuppressive mechanism of FK-506. In our ongoing search for superior immunosuppressive agents, we have synthesized several analogues of FK-506 and tested their mechanistic and immunosuppressive actions. It was found that C-18 hydroxyl analogues of ascomycin, an analogue of FK-506 also called FR900520, bound tightly to immunophilin FKBP-12, but do not show any immunosuppressive activity in vitro or in vivo despite good bioavailability. Further, they reverse the inhibition of calcineurin caused by FK-506/FKBP-12 complex.