Merck
  • Engineered Au Core@Prussian Blue Analogous Shell Nanoheterostructures: Their Magnetic and Optical Properties.
Engineered Au Core@Prussian Blue Analogous Shell Nanoheterostructures: Their Magnetic and Optical Properties.
Chemistry (Weinheim an der Bergstrasse, Germany) (2017-02-12)
Guillaume Maurin-Pasturel, Jérôme Long, Maria A Palacios, Christian Guérin, Clarence Charnay, Marc-Georg Willinger, Alexander A Trifonov, Joulia Larionova, Yannick Guari
ABSTRACT
We report a new approach for the synthesis of multifunctional Au core@Prussian Blue analogous (PBA) shell nanoheterostructures that involves PBA shell growth on the surface of cyanide-stabilized gold nanoparticles. It permits the assembly of Au@KNi