Merck
Inhibition of HSF1 suppresses the growth of hepatocarcinoma cell lines
Oncotarget (2017-09-15)
Antonio Cigliano, Chunmei Wang, Maria G Pilo, Marta Szydlowska, Stefania Brozzetti, Gavinella Latte, Giovanni M Pes, Rosa M Pascale, Maria A Seddaiu, Gianpaolo Vidili, Silvia Ribback, Frank Dombrowski, Matthias Evert, Xin Chen, Diego F Calvisi
ABSTRACT
Upregulation of the heat shock transcription factor 1 (HSF1) has been described as a frequent event in many cancer types, but its oncogenic role in hepatocellular carcinoma (HCC) remains poorly delineated. In the present study, we assessed the function(s) of HSF1 in hepatocarcinogenesis via
Sigma-Aldrich
MISSION® esiRNA, targeting human HSF1