Merck
  • Formulation by design based risperidone nano soft lipid vesicle as a new strategy for enhanced transdermal drug delivery: In-vitro characterization, and in-vivo appraisal.
Formulation by design based risperidone nano soft lipid vesicle as a new strategy for enhanced transdermal drug delivery: In-vitro characterization, and in-vivo appraisal.
Materials science & engineering. C, Materials for biological applications (2017-04-19)
Syed Sarim Imam, Abdul Ahad, Mohammed Aqil, Mohd Akhtar, Yasmin Sultana, Asgar Ali
ABSTRACT
The present study was designed to formulate and optimize transdermal risperidone soft lipid vesicles. The formulation optimized with phospholipid, safranal and ethanol were incorporated as permeation and absorption enhancers. The optimized risperidone soft lipid vesicle was further evaluated for skin irritation study, in-vivo pharmacokinetic study and locomotor activity. Three factor three level Box-Behnken design (BBD) was used to statistically optimize soft lipid vesicle using safranal (A), ethanol (B)and phospholipid (C) as independent variable, while their effect was observed for vesicle size (Y
Sigma-Aldrich
Safranal, ≥90%, stabilized