Merck
Metabolomic analysis shows differential hepatic effects of T
Scientific reports (2017-05-19)
Liliana F Iannucci, Federica Cioffi, Rosalba Senese, Fernando Goglia, Antonia Lanni, Paul M Yen, Rohit A Sinha
ABSTRACT
Nonalcoholic fatty liver disease (NAFLD) is a major health problem worldwide, and is often associated with lipotoxic injury, defective mitochondrial function, and insulin resistance. Thyroid hormones (THs) are important regulators of hepatic lipid metabolism. Among the THs, diiodothyronine (T
Sigma-Aldrich
3,3′-Diiodo-L-thyronine (T2) hydrochloride, 98% (CP)