Merck
  • Success of murine embryo transfer increased by a modified transfer pipette.
Success of murine embryo transfer increased by a modified transfer pipette.
The Journal of reproduction and development (2008-11-22)
Zheng Zhang, Xiaoyan Lv, Yidong Wang, Ye Chen, Rong Zheng, Huan Sun, Guohui Bian, Yan Xiao, Qingwei Li, Qiutan Yang, Jianzhong Ai, Jingjing Duan, Ruizhi Tan, Yuhang Liu, Yang Yang, Yuquan Wei, Qin Zhou
ABSTRACT
Embryos transfer is one of the most critical steps for generating genetically modified mice. Because of the limitations of the current transfer equipment and techniques, such as discharge of freshly transferred embryos and the necessity of sophisticated transfer skills, the success ratio of implantation may not well satisfy the requirements for mass production of high quality animals in the field of genetically modified mice. In this study, we describe a modified embryo transfer pipette with a syringe-like tip that can easily be applied to mouse embryo transfer. Using this improved method, we show that the procedures for mouse embryo implantation are simplified and easier to perform; moreover, the viability rate of mouse embryos is 20% higher than that achieved with conventional methods. Our modified tool and improved transfer technique are effective, time-saving and less invasive, resulting in increased success of embryo transfer.
Sigma-Aldrich
Chorionic gonadotropin human, lyophilized powder, suitable for mouse embryo cell culture