Merck
  • Chondroitin sulphate A (CSA)-binding of single recombinant Duffy-binding-like domains is not restricted to Plasmodium falciparum Erythrocyte Membrane Protein 1 expressed by CSA-binding parasites.
Chondroitin sulphate A (CSA)-binding of single recombinant Duffy-binding-like domains is not restricted to Plasmodium falciparum Erythrocyte Membrane Protein 1 expressed by CSA-binding parasites.
International journal for parasitology (2009-03-28)
Mafalda Resende, Sisse B Ditlev, Morten A Nielsen, Sabrina Bodevin, Silas Bruun, Vera V Pinto, Henrik Clausen, Louise Turner, Thor G Theander, Ali Salanti, Madeleine Dahlbäck
ABSTRACT
Individuals living in areas with high Plasmodium falciparum transmission acquire immunity to malaria over time and adults have a markedly reduced risk of contracting severe disease. However, pregnant women constitute an important exception. Pregnancy-associated malaria is a major cause of mother and offspring morbidity, such as severe maternal anaemia and low birth-weight, and is characterised by selective accumulation of parasite-infected erythrocytes (IE) in the placenta. A P. falciparum protein named VAR2CSA, which belongs to the large P. falciparum Erythrocyte Membrane Protein 1 (PfEMP1) family, enables the IE to bind chondroitin sulphate A (CSA) in the placenta. Knock-out studies have demonstrated the exclusive capacity of VAR2CSA to mediate IE binding to CSA, and it has been shown that four of the six Duffy-binding-like (DBL) domains of VAR2CSA have the ability to bind CSA in vitro. In this study, we confirm the CSA-binding of these DBL domains, however, the analysis of a number of DBL domains of a non-VAR2CSA origin shows that CSA-binding is not exclusively restricted to VAR2CSA DBL domains. Furthermore, we show that the VAR2CSA DBL domains as well as other DBL domains also bind heparan sulphate. These data explain a number of publications describing CSA-binding domains derived from PfEMP1 antigens not involved in placental adhesion. The data suggest that the ability of single domains to bind CSA does not predict the functional capacity of the whole PfEMP1 and raises doubt whether the CSA-binding domains of native VAR2CSA have been correctly identified.