Merck
  • The impact of wine components on fractionation of Cu and Fe in model wine systems: Macromolecules, phenolic and sulfur compounds.
The impact of wine components on fractionation of Cu and Fe in model wine systems: Macromolecules, phenolic and sulfur compounds.
Food research international (Ottawa, Ont.) (2017-06-15)
Nikolaos Kontoudakis, Mark Smith, Anque Guo, Paul A Smith, Geoffrey R Scollary, Eric N Wilkes, Andrew C Clark
ABSTRACT
A variety of techniques have been developed with the ability to measure different forms of metals in wine with the ultimate aim of providing a more accurate indicator of metal induced spoilage of wine. This study was conducted in order to identify which wine components influence the measurement of Cu and Fe in their fractionated and/or electrochemically active forms. The measurement techniques involved detection of labile Cu by stripping potentiometry and fractionation of Cu and Fe by sequential solid phase extraction, with detection by inductively coupled plasma-optical emission spectroscopy. The wine components assessed included those extracted from wine (red wine tannin, white wine protein, white wine polysaccharide, red wine polyphenol, white wine polyphenol), and commercially available monomeric compounds, including phenolic compounds and sulfur-containing compounds. For Cu, only hydrogen sulfide, which is known to induce the formation of Cu(I) sulfide, showed any appreciable influence on the fractionation and electrochemical detection of Cu. This form of Cu was also identified as the major component of red and white wines. For Fe, the fractionation was different for red versus white wine, and influenced significantly by extracted red wine polyphenol, (-)-epicatechin, gallic acid and tartaric acid. The wine components showed more influence on Fe at pH4.00 compared to pH3.25. These results enable a targeted use of these techniques in the assessment of metal-induced spoilage of wine.
Sigma-Aldrich
Sodium ethanethiolate, technical, ~90% (RT)
Sigma-Aldrich
Copper(II) sulfate pentahydrate, LR, ≥99%
Sigma-Aldrich
Sodium methanethiolate, technical, ≥90% (RT)