Merck
  • A Robust, Water-Based, Functional Binder Framework for High-Energy Lithium-Sulfur Batteries.
A Robust, Water-Based, Functional Binder Framework for High-Energy Lithium-Sulfur Batteries.
ChemSusChem (2017-05-26)
Matthew J Lacey, Viking Österlund, Andreas Bergfelt, Fabian Jeschull, Tim Bowden, Daniel Brandell
ABSTRACT
We report here a water-based functional binder framework for the lithium-sulfur battery systems, based on the general combination of a polyether and an amide-containing polymer. These binders are applied to positive electrodes optimised towards high-energy electrochemical performance based only on commercially available materials. Electrodes with up to 4 mAh cm
Sigma-Aldrich
N,N-Dimethylacrylamide, 99%, contains 500 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
1-(2-Hydroxyethyl)-2-pyrrolidone, 98%