Merck
  • On the use of the experimentally determined enzyme inhibition constant as a measure of absolute binding affinity.
On the use of the experimentally determined enzyme inhibition constant as a measure of absolute binding affinity.
Biochemical and biophysical research communications (2017-06-03)
Fouad H Darras, Yuan-Ping Pang
ABSTRACT
Defined as a state function representing an inhibitor's absolute affinity for its target enzyme, the experimentally determined enzyme inhibition constant (K
Sigma-Aldrich
Acetylthiocholine chloride, ≥99% (TLC), powder
Sigma-Aldrich
Acetylcholinesterase from Electrophorus electricus (electric eel), Type V-S, lyophilized powder, ≥1,000 units/mg protein