Merck
  • Drug- and Drug Abuse-Associated Hyperbilirubinemia: Experience With Atazanavir.
Drug- and Drug Abuse-Associated Hyperbilirubinemia: Experience With Atazanavir.
Clinical pharmacology in drug development (2017-03-07)
Jayanta Roy-Chowdhury, Namita Roy-Chowdhury, Irving Listowsky, Allan W Wolkoff
ABSTRACT
Hyperbilirubinemia is a common finding in individuals with a history of substance abuse. Although this may indicate a serious disorder of liver function, this is not always the case. An understanding of bilirubin formation, metabolism, and transport can provide a helpful approach to dealing with these patients. This is typified by studies of patients treated with the antiretroviral drug atazanavir. Atazanavir has been associated with hyperbilirubinemia in as many as one-third of individuals for whom it has been prescribed, evoking concerns of hepatotoxicity. The studies in this report were designed to determine mechanisms by which this occurs. The data show that this drug inhibits the enzyme UDP-glucuronosyl transferase-1A1, responsible for conjugating bilirubin with glucuronic acid. This conjugation step is required for bilirubin excretion into bile, and when it is inhibited, bilirubin refluxes from the liver into the circulation, causing unconjugated hyperbilirubinemia. Other parameters of bilirubin formation, binding to albumin in the circulation, uptake into hepatocytes, and intracellular protein binding in hepatocytes were unaffected by atazanavir. The effect of atazanavir on serum bilirubin levels is reversible, consistent with lack of structural damage to the liver.
Sigma-Aldrich
Atazanavir, ≥98% (HPLC)
Pricing and availability is not currently available.