Merck
  • Centromeres are maintained by fastening CENP-A to DNA and directing an arginine anchor-dependent nucleosome transition.
Centromeres are maintained by fastening CENP-A to DNA and directing an arginine anchor-dependent nucleosome transition.
Nature communications (2017-06-10)
Lucie Y Guo, Praveen Kumar Allu, Levani Zandarashvili, Kara L McKinley, Nikolina Sekulic, Jennine M Dawicki-McKenna, Daniele Fachinetti, Glennis A Logsdon, Ryan M Jamiolkowski, Don W Cleveland, Iain M Cheeseman, Ben E Black
ABSTRACT
Maintaining centromere identity relies upon the persistence of the epigenetic mark provided by the histone H3 variant, centromere protein A (CENP-A), but the molecular mechanisms that underlie its remarkable stability remain unclear. Here, we define the contributions of each of the three candidate CENP-A nucleosome-binding domains (two on CENP-C and one on CENP-N) to CENP-A stability using gene replacement and rapid protein degradation. Surprisingly, the most conserved domain, the CENP-C motif, is dispensable. Instead, the stability is conferred by the unfolded central domain of CENP-C and the folded N-terminal domain of CENP-N that becomes rigidified 1,000-fold upon crossbridging CENP-A and its adjacent nucleosomal DNA. Disrupting the 'arginine anchor' on CENP-C for the nucleosomal acidic patch disrupts the CENP-A nucleosome structural transition and removes CENP-A nucleosomes from centromeres. CENP-A nucleosome retention at centromeres requires a core centromeric nucleosome complex where CENP-C clamps down a stable nucleosome conformation and CENP-N fastens CENP-A to the DNA.
Sigma-Aldrich
MISSION® esiRNA, targeting human GAPDH
Sigma-Aldrich
MISSION® esiRNA, targeting human CENPN