Merck
  • Scaffold-free cartilage tissue engineering with a small population of human nasoseptal chondrocytes.
Scaffold-free cartilage tissue engineering with a small population of human nasoseptal chondrocytes.
The Laryngoscope (2016-11-20)
Loraine L Y Chiu, William T H To, John M Lee, Stephen D Waldman
ABSTRACT
Cartilage tissue engineering is a promising approach to provide suitable materials for nasal reconstruction; however, it typically requires large numbers of cells. We have previously shown that a small number of chondrocytes cultivated within a continuous flow bioreactor can elicit substantial tissue growth, but translation to human chondrocytes is not trivial. Here, we aimed to demonstrate the application of the bioreactor to generate large-sized tissues from a small population of primary human nasoseptal chondrocytes. Experimental study. Chondrocytes were cultured in the bioreactor using different medium compositions, with varying amounts of serum and with or without growth factors. Resulting engineered tissues were analyzed for physical properties, biochemical composition, tissue microstructure, and protein localization. Bioreactor-cultivated constructs grown with serum and growth factors (basic fibroblast growth factor and transforming growth factor beta 2) had greater thickness, as well as DNA and glycosaminoglycan (GAG) contents, compared to low serum and no growth factor controls. These constructs also showed the most intense proteoglycan and collagen II staining. The combination of bioreactor conditions, serum, and growth factors allowed the generation of large, thick scaffold-free human cartilaginous tissues that resembled the native nasoseptal cartilage. There also may be implications for patient selection in future clinical applications of these engineered tissues because their GAG content decreased with donor age. NA. Laryngoscope, 127:E91-E99, 2017.
Sigma-Aldrich
ITS Liquid Media Supplement (100×), liquid, sterile-filtered, BioReagent, suitable for cell culture