- Interaction of organic ions with proteins.
Interaction of organic ions with proteins.
Soft matter (2017-01-18)
Leonor Pérez-Fuentes, Carlos Drummond, Jordi Faraudo, Delfi Bastos-González
PMID28093583
ABSTRACT
In this study we have investigated how different proteins interact with big organic ions. Two ions that are similar in size and chemical structure (Ph
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, fatty acid free, essentially globulin free, pH 7, ≥98%