Merck
  • A simple synthetic route to polyoxovanadate-based organic-inorganic hybrids using EEDQ as an ester coupling agent.
A simple synthetic route to polyoxovanadate-based organic-inorganic hybrids using EEDQ as an ester coupling agent.
Dalton transactions (Cambridge, England : 2003) (2017-03-23)
Aruuhan Bayaguud, JianDa Li, Shan She, Yongge Wei
ABSTRACT
A reaction strategy for the post-functionalization of hexavanadate derivatives is presented herein. In this study, five polyoxovanadate-based organic-inorganic hybrids TBA
Sigma-Aldrich
1,12-Dodecanediol, 99%