Merck
Enantiospecific Allosteric Modulation of Cannabinoid 1 Receptor.
ACS chemical neuroscience (2017-01-20)
Robert B Laprairie, Pushkar M Kulkarni, Jeffrey R Deschamps, Melanie E M Kelly, David R Janero, Maria G Cascio, Lesley A Stevenson, Roger G Pertwee, Terrence P Kenakin, Eileen M Denovan-Wright, Ganesh A Thakur
ABSTRACT
The cannabinoid 1 receptor (CB1R) is one of the most widely expressed metabotropic G protein-coupled receptors in brain, and its participation in various (patho)physiological processes has made CB1R activation a viable therapeutic modality. Adverse psychotropic effects limit the clinical utility of CB1R orthosteric agonists and have promoted the search for CB1R positive allosteric modulators (PAMs) with the promise of improved drug-like pharmacology and enhanced safety over typical CB1R agonists. In this study, we describe the synthesis and in vitro and ex vivo pharmacology of the novel allosteric CB1R modulator GAT211 (racemic) and its resolved enantiomers, GAT228 (R) and GAT229 (S). GAT211 engages CB1R allosteric site(s), enhances the binding of the orthosteric full agonist [
Sigma-Aldrich
GAT229, ≥98% (HPLC)
Sigma-Aldrich
GAT228, ≥98% (HPLC)