Merck
  • CXCL5 Plays a Promoting Role in Osteosarcoma Cell Migration and Invasion in Autocrine- and Paracrine-Dependent Manners.
CXCL5 Plays a Promoting Role in Osteosarcoma Cell Migration and Invasion in Autocrine- and Paracrine-Dependent Manners.
Oncology research (2017-03-10)
Hongsheng Dang, Wuzhou Wu, Bo Wang, Cao Cui, Juwei Niu, Jie Chen, Ziqiu Chen, Yi Liu
ABSTRACT
CXCL5, a CXC-type chemokine, is an important attractant for granulocytic immune cells by binding to its receptor CXCR2. Recently, CXCL5/CXCR2 has been found to play an oncogenic role in many human cancers. However, the exact role of CXCL5 in osteosarcoma cell migration and invasion has not been revealed. Here we found that the protein expression of CXCL5 was significantly increased in osteosarcoma tissues compared with that in matched adjacent nontumor tissues. Moreover, the expression of CXCL5 was significantly associated with advanced clinical stage and metastasis. Further investigation showed that the CXCL5 expression levels were also significantly increased in osteosarcoma cell lines, including Saos-2, MG63, U2OS, and SW1353, when compared with those in normal osteoblast hFoB1.19 cells. U2OS cells were further transfected with CXCL5-specific siRNA or overexpression plasmid. Knockdown of CXCL5 significantly suppressed U2OS cell migration and invasion. On the contrary, overexpression of CXLC5 remarkably promoted the migration and invasion of U2OS cells. Interestingly, both exogenous CXCL5 treatment and the conditioned medium of CXCL5-overexpressing hFoB1.19 cells could also enhance the migration and invasion of U2OS cells, suggesting that the promoting role of CXCL5 in U2OS cell migration and invasion is also in a paracrine-dependent manner. According to these data, our study demonstrates that CXCL5 is upregulated in osteosarcoma and may play an oncogenic role in osteosarcoma metastasis. Therefore, CXCL5 may become a potential therapeutic target for osteosarcoma treatment.
Sigma-Aldrich
MISSION® esiRNA, targeting human CXCL5