Merck
  • Phospholipase D1 deficiency in mice causes nonalcoholic fatty liver disease via an autophagy defect.
Phospholipase D1 deficiency in mice causes nonalcoholic fatty liver disease via an autophagy defect.
Scientific reports (2016-12-16)
Jang Ho Hur, Shi-Young Park, Claudia Dall'Armi, Jae Sung Lee, Gilbert Di Paolo, Hui-Young Lee, Mee-Sup Yoon, Do Sik Min, Cheol Soo Choi
ABSTRACT
Nonalcoholic fatty liver disease (NAFLD) is characterized by the accumulation of triglycerides (TG) as lipid droplets in the liver. Although lipid-metabolizing enzymes are considered important in NAFLD, the involvement of phospholipase D1 (PLD1) has not yet been studied. Here, we show that the genetic ablation of PLD1 in mice induces NAFLD due to an autophagy defect. PLD1 expression was decreased in high-fat diet-induced NAFLD. Subsequently, PLD1 deficiency led to an increase in hepatic TGs and liver weight. Autophagic flux was blocked in Pld1-/- hepatocytes, with decreased β-oxidation rate, reduced oxidation-related gene expression, and swollen mitochondria. The dynamics of autophagy was restored by treatment with the PLD product, phosphatidic acid (PA) or adenoviral PLD1 expression in Pld1-/- hepatocytes, confirming that lysosomal PA produced by PLD1 regulates autophagy. Notably, PLD1 expression in Pld1-/- liver significantly reduced hepatic lipid accumulation, compared with Pld1-/- liver. Thus, PLD1 plays an important role in hepatic steatosis via the regulation of autophagy.
Sigma-Aldrich
Medium 199, With Earle′s salts, L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Phospholipase D Activity Assay Kit, sufficient for 100 colorimetric or fluorometric tests
Sigma-Aldrich
VU0155069, ≥98% (HPLC)