Merck
  • The multi-target drug BAI induces apoptosis in various human cancer cells through modulation of Bcl-xL protein.
The multi-target drug BAI induces apoptosis in various human cancer cells through modulation of Bcl-xL protein.
International journal of oncology (2016-11-15)
Shin Kim, Dong Eun Kim, Taeg Kyu Kwon, Jinho Lee, Jong-Wook Park
ABSTRACT
Previous studies have demonstrated the anticancer effects of the newly developed cyclin-dependent kinase inhibitor BAI in various cancer cells. However, the molecular mechanisms of the cellular effects induced by BAI have not been fully elucidated. The objective of this study was to investigate the mechanisms underlying the regulation of B cell lymphoma-2 (Bcl-2) family proteins in BAI-induced apoptosis of cancer cells. BAI induced poly(ADP-ribose) polymerase cleavage and DEVDase activation dose- and time-dependently. However, BAI-induced apoptosis was not involved in reactive oxygen species generation or mitogen-activated protein kinases pathways. On the other hand, BAI reduced the mitochondrial membrane potential (∆ψm) dose- and time-dependently, and induced the release of apoptosis-inducing factor (AIF) and cytochrome c from mitochondria in A549 and Caki cells. Furthermore, BAI-induced apoptosis was strongly associated with downregulation of B-cell lymphoma-extra large (Bcl-xL), but not Bcl-2, and BAI modulated the interactions among p53 and Bcl-2 family proteins in human cancer cells. Taken together, these results revealed that the regulations of Bcl-2 family proteins are correlated with BAI-induced apoptosis, suggesting that BAI is a potential multi-target agent of cancer.
Sigma-Aldrich
1,1′-(Methylenedi-4,1-phenylene)bismaleimide, 95%