Merck
  • Dying glioma cells establish a proangiogenic microenvironment through a caspase 3 dependent mechanism.
Dying glioma cells establish a proangiogenic microenvironment through a caspase 3 dependent mechanism.
Cancer letters (2016-11-09)
Xiao Feng, Yang Yu, Sijia He, Jin Cheng, Yanping Gong, Zhengxiang Zhang, Xuguang Yang, Bing Xu, Xinjian Liu, Chuan-Yuan Li, Ling Tian, Qian Huang
ABSTRACT
Vascular recovery or re-angiogenesis after radiotherapy plays a significant role in tumor recurrence, whereas molecular mechanisms of this process remain elusive. In this work, we found that dying glioma cells promoted post-irradiation angiogenesis through a caspase 3 dependent mechanism. Evidence in vitro and in vivo indicated that caspase 3 inhibition undermined proangiogenic effects of dying glioma cells. Proteolytic inactivation of caspase 3 in glioma cells reduced tumorigenicity. Importantly, we identified that NF-κB/COX-2/PGE2 axis acted as downstream signaling of caspase 3, mediating proangiogenic response after irradiation. Additionally, VEGF-A, regulated by caspase 3 possibly through phosphorylated eIF4E, was recognized as another downstream factor participating in the proangiogenic response. In conclusion, these data demonstrated that caspase 3 in dying glioma cells supported the proangiogenic response after irradiation by governing NF-κB/COX-2/PGE2 axis and p-eIF4E/VEGF-A signaling. While inducing caspase 3 activation has been a generally-adopted notion in cancer therapeutics, our study counterintuitively illustrated that caspase 3 activation in dying glioma cells unfavorably supported post-irradiation angiogenesis. This double-edged role of caspase 3 suggested that taming caspase 3 from the opposite side, not always activating it, may provide novel therapeutic strategies due to restricted post-irradiation angiogenesis.
Sigma-Aldrich
Prostaglandin B2, ≥98%, synthetic
Pricing and availability is not currently available.