Merck
  • The Planorbid Snail Biomphalaria glabrata Expresses a Hemocyanin-Like Sequence in the Albumen Gland.
The Planorbid Snail Biomphalaria glabrata Expresses a Hemocyanin-Like Sequence in the Albumen Gland.
PloS one (2016-12-31)
Janeth J Peña, Coen M Adema
ABSTRACT
The parasitic flatworm Schistosoma mansoni, causative agent of human intestinal schistosomiasis in South America, relies importantly on the freshwater snail Biomphalaria glabrata as intermediate host to achieve development of cercariae that infect humans. The recommendation from the World Health Organization (WHO) to integrate snail control in efforts to counter schistosomiasis transmission provides impetus for in depth study of B. glabrata biology. Our analysis indicates that two distinct hemocyanin-like genes (hcl-1 and hcl-2) are present in B. glabrata, a snail that uses hemoglobin for oxygen transport. Characterization of BAC clones yielded the full length hcl-1 gene, which is comprised of three functional unit (FU) domains at the amino acid level. Database searches and in silico analyses identified the second hcl gene (hcl-2), composed of six FU domains. Both genes are unusual for lacking canonical residues and having fewer FU domains than typical molluscan hemocyanins that contain 7-8 FUs. Reverse transcription PCR demonstrated that Hcl-1 is expressed in a manner that correlates with reproductive maturity in the albumen gland (AG), an immune- and reproduction-relevant organ. Immune cross-reactivity with anti-keyhole limpet hemocyanin (α-KLH) antiserum and tandem-mass spectrometry validated the presence of Hcl-1 protein in the AG and egg mass fluid (EMF). The evolutionary conservation of hemocyanin-like sequences in B. glabrata in the presence of the oxygen carrier hemoglobin, combined with our results, suggest that the Hcl-1protein has a functional role in general and/or reproductive biology. Further investigations are needed to explore Hcl-1 as a potential target for snail control.