Merck
  • Cyclin A is redundant in fibroblasts but essential in hematopoietic and embryonic stem cells.
Cyclin A is redundant in fibroblasts but essential in hematopoietic and embryonic stem cells.
Cell (2009-07-14)
Ilona Kalaszczynska, Yan Geng, Tadafumi Iino, Shin-ichi Mizuno, Yoon Choi, Ilona Kondratiuk, Daniel P Silver, Debra J Wolgemuth, Koichi Akashi, Piotr Sicinski
ABSTRACT
Cyclins are regulatory subunits of cyclin-dependent kinases. Cyclin A, the first cyclin ever cloned, is thought to be an essential component of the cell-cycle engine. Mammalian cells encode two A-type cyclins, testis-specific cyclin A1 and ubiquitously expressed cyclin A2. Here, we tested the requirement for cyclin A function using conditional knockout mice lacking both A-type cyclins. We found that acute ablation of cyclin A in fibroblasts did not affect cell proliferation, but led to prolonged expression of another cyclin, cyclin E, across the cell cycle. However, combined ablation of all A- and E-type cyclins extinguished cell division. In contrast, cyclin A function was essential for cell-cycle progression of hematopoietic and embryonic stem cells. Expression of cyclin A is particularly high in these compartments, which might render stem cells dependent on cyclin A, whereas in fibroblasts cyclins A and E play redundant roles in cell proliferation.