Merck
CO2-inhibited neurons in the medullary raphé are GABAergic.
Respiratory physiology & neurobiology (2014-08-05)
Kimberly E Iceman, Andrea E Corcoran, Barbara E Taylor, Michael B Harris
ABSTRACT
Previous studies have reported subsets of medullary raphé neurons that are either stimulated or inhibited by CO2/pH in vitro, in situ, and in vivo. We tested the hypothesis that medullary raphé CO2-inhibited neurons are GABAergic. Extracellular recordings in unanesthetized juvenile in situ rat preparations showed reversible hypercapnia-induced suppression of 19% (63/323) of medullary raphé neurons, and this suppression persisted after antagonism of NMDA, AMPA/kainate, and GABAA receptors. We stained a subset of CO2-inhibited cells and found that most (11/12) had glutamic acid decarboxylase 67 immunoreactivity (GAD67-ir). These data indicate that the majority of acidosis-inhibited medullary raphé neurons are GABAergic, and that their chemosensitivity is independent of major fast synaptic inputs. Thus, CO2-sensitive GABAergic neurons may play a role in central CO2/pH chemoreception.
Sigma-Aldrich
Anti-GAD67 Antibody, clone 1G10.2, clone 1G10.2, Chemicon®, from mouse