Merck
  • The green microalga Tetraselmis suecica reduces oxidative stress and induces repairing mechanisms in human cells.
The green microalga Tetraselmis suecica reduces oxidative stress and induces repairing mechanisms in human cells.
Scientific reports (2017-01-25)
Clementina Sansone, Christian Galasso, Ida Orefice, Genoveffa Nuzzo, Elvira Luongo, Adele Cutignano, Giovanna Romano, Christophe Brunet, Angelo Fontana, Francesco Esposito, Adrianna Ianora
ABSTRACT
Green microalgae contain many active pigments such as carotenoids having antioxidant and protective activity on human cells. Here we investigate the biological activity of an ethanol/water extract of the marine green microalga Tetraselmis suecica containing high levels of carotenoids such as the xanthophylls lutein, violaxanthin, neoxanthin, antheraxanthin and loroxanthin esters. This extract has a strong antioxidant and repairing activity in the human lung cancer cell line (A549) as shown by the increased expression of dehydrocholesterol reductase-24 (DHCR24) and prostaglandin reductase 1 (PTGR1) genes and proteins. The extract also reduces prostaglandin E2 (PGE2) levels in cells damaged by H2O2 and has tissue repairing effects on reconstructed human epidermal tissue cells (EpiDermTM) indicating a potential cosmeceutical activity of this microalgal species.
Sigma-Aldrich
Anti-GPX4 (Isoform A and C) antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially IgG-free, low endotoxin, BioReagent, suitable for cell culture
Sigma-Aldrich
Desmosterol, ≥84% (GC)