Merck
  • Preservation of Capillary-beds in Rat Lung Tissue Using Optimized Chemical Decellularization.
Preservation of Capillary-beds in Rat Lung Tissue Using Optimized Chemical Decellularization.
Journal of materials chemistry. B (2013-10-02)
Ryan J Nagao, Yafei Ouyang, Renee Keller, Curtis Lee, Laura J Suggs, Christine E Schmidt
ABSTRACT
Promoting regeneration using scaffolds created by decellularizing native tissue is becoming a popular technique applied to a variety of tissues. We demonstrate a method to decellularize highly vascular tissue keeping the vascular structure intact down to the capillary scale. Using vascular corrosion casting (VCC), we created a method for quantitatively assessing the functionality of vascular extracellular matrix (ECM) following decellularization. Murine lung tissue was decellularized using a number of techniques, then characterized using standard histological methods, as well as our quantitative VCC (qVCC) technique. Using an optimized acellular method, we successfully decellularized lung tissue while leaving behind a patent vascular network based on qualitative and quantitative histological methods.