Merck
  • A simplified calculation procedure for mass isotopomer distribution analysis (MIDA) based on multiple linear regression.
A simplified calculation procedure for mass isotopomer distribution analysis (MIDA) based on multiple linear regression.
Journal of mass spectrometry : JMS (2016-07-09)
Mario Fernández-Fernández, Pablo Rodríguez-González, J Ignacio García Alonso
ABSTRACT
We have developed a novel, rapid and easy calculation procedure for Mass Isotopomer Distribution Analysis based on multiple linear regression which allows the simultaneous calculation of the precursor pool enrichment and the fraction of newly synthesized labelled proteins (fractional synthesis) using linear algebra. To test this approach, we used the peptide RGGGLK as a model tryptic peptide containing three subunits of glycine. We selected glycine labelled in two
Sigma-Aldrich
Fmoc-Gly-OH, ≥98.0% (T)