Merck
  • Copper-carbene complexes as catalysts in the synthesis of functionalized styrenes and aliphatic alkenes.
Copper-carbene complexes as catalysts in the synthesis of functionalized styrenes and aliphatic alkenes.
The Journal of organic chemistry (2006-12-30)
Hélène Lebel, Michaël Davi, Silvia Díez-Gonzalez, Steven P Nolan
ABSTRACT
(NHC)-Cu (NHC = N-heterocyclic carbene) complexes efficiently catalyzed the methylenation of a variety of aliphatic and aromatic aldehydes and ketones in the presence of trimethylsilyldiazomethane, triphenylphosphine, and 2-propanol. The copper catalysts are not only inexpensive compared to rhodium complexes, but they also exhibit better functional group compatibility with aromatic aldehydes and ketones. Indeed very high yields were obtained for the formation of styrenes containing nitro, trifluoromethyl, amino, and ester groups, as well as for pyridine-, pyrrole-, and indole-substituted alkenes.
Sigma-Aldrich
Chloro[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene]copper(I)