Merck
  • Oxidized LDL induces FAK-dependent RSK signaling to drive NF-κB activation and VCAM-1 expression.
Oxidized LDL induces FAK-dependent RSK signaling to drive NF-κB activation and VCAM-1 expression.
Journal of cell science (2016-02-26)
Arif Yurdagul, Florian J Sulzmaier, Xiao L Chen, Christopher B Pattillo, David D Schlaepfer, A Wayne Orr
ABSTRACT
Oxidized low-density lipoprotein (oxLDL) accumulates early in atherosclerosis and promotes endothelial nuclear factor κB (NF-κB) activation, proinflammatory gene expression and monocyte adhesion. Like for other atherogenic factors, oxLDL-induced proinflammatory responses requires integrin-dependent focal adhesion kinase (FAK, also known as PTK2) signaling; however, the mechanism by which FAK mediates oxLDL-dependent NF-κB signaling has yet to be revealed. We now show that oxLDL induces NF-κB activation and VCAM-1 expression through FAK-dependent IκB kinase β (IKKβ, also known as IKBKB) activation. We further identify FAK-dependent activation of p90 ribosomal S6 kinase family proteins (RSK) as a crucial mediator of oxLDL-dependent IKKβ and NF-κB signaling, as inhibiting RSK blocks oxLDL-induced IKKβ and NF-κB activation, VCAM-1 expression and monocyte adhesion. Finally, transgenic mice containing a kinase-dead mutation in FAK specifically in the endothelial cells show reduced RSK activity, decreased VCAM-1 expression and reduced macrophage accumulation in regions of early atherosclerosis. Taken together, our data elucidates a new mechanism whereby oxLDL-induced endothelial FAK signaling drives an ERK-RSK pathway to activate IKKβ and NF-κB signaling and proinflammatory gene expression.