- Generation of iPSC lines from primary human chorionic villi cells.
Generation of iPSC lines from primary human chorionic villi cells.
Stem cell research (2016-03-19)
Björn Lichtner, Peggy Matz, James Adjaye
PMID26987929
ABSTRACT
Primary human chorionic villi (CV) cells were used to generate the iPSC line by retroviral transduction of the four Yamanaka-factors OCT4, SOX2, KLF4 and c-MYC. Pluripotency was confirmed both in vivo and in vitro. The transcriptomes of the CV-derived iPSC lines and the human embryonic stem cell lines-H1 and H9 have a Pearson correlation of 0.929 and 0.943 respectively.
Sigma-Aldrich
Monoclonal Anti-AFP antibody produced in mouse, clone 1G7, purified immunoglobulin, buffered aqueous solution