Merck
  • Microtubule minus-end binding protein CAMSAP2 controls axon specification and dendrite development.
Microtubule minus-end binding protein CAMSAP2 controls axon specification and dendrite development.
Neuron (2014-06-09)
Kah Wai Yau, Sam F B van Beuningen, Inês Cunha-Ferreira, Bas M C Cloin, Eljo Y van Battum, Lena Will, Philipp Schätzle, Roderick P Tas, Jaap van Krugten, Eugene A Katrukha, Kai Jiang, Phebe S Wulf, Marina Mikhaylova, Martin Harterink, R Jeroen Pasterkamp, Anna Akhmanova, Lukas C Kapitein, Casper C Hoogenraad
ABSTRACT
In neurons, most microtubules are not associated with a central microtubule-organizing center (MTOC), and therefore, both the minus and plus-ends of these non-centrosomal microtubules are found throughout the cell. Microtubule plus-ends are well established as dynamic regulatory sites in numerous processes, but the role of microtubule minus-ends has remained poorly understood. Using live-cell imaging, high-resolution microscopy, and laser-based microsurgery techniques, we show that the CAMSAP/Nezha/Patronin family protein CAMSAP2 specifically localizes to non-centrosomal microtubule minus-ends and is required for proper microtubule organization in neurons. CAMSAP2 stabilizes non-centrosomal microtubules and is required for neuronal polarity, axon specification, and dendritic branch formation in vitro and in vivo. Furthermore, we found that non-centrosomal microtubules in dendrites are largely generated by γ-Tubulin-dependent nucleation. We propose a two-step model in which γ-Tubulin initiates the formation of non-centrosomal microtubules and CAMSAP2 stabilizes the free microtubule minus-ends in order to control neuronal polarity and development.