Merck
  • Effects of immune supplementation and immune challenge on oxidative status and physiology in a model bird: implications for ecologists.
Effects of immune supplementation and immune challenge on oxidative status and physiology in a model bird: implications for ecologists.
The Journal of experimental biology (2010-10-05)
Janske van de Crommenacker, Nicholas P C Horrocks, Maaike A Versteegh, Jan Komdeur, B Irene Tieleman, Kevin D Matson
ABSTRACT
One route to gain insight into the causes and consequences of ecological differentiation is to understand the underlying physiological mechanisms. We explored the relationships between immunological and oxidative status and investigated how birds cope physiologically with the effects of immune-derived oxidative damage. We successively implemented two experimental manipulations to alter physiological status in a model bird species: the homing pigeon (Columba livia). The first manipulation, an immune supplementation, was achieved by oral administration of lysozyme, a naturally occurring and non-specific antimicrobial enzyme. The second manipulation, an immune challenge, took the form of an injection with lipopolysaccharide, a bacterial endotoxin. Between groups of lysozyme-treated and control birds, we compared lipopolysaccharide-induced changes in reactive oxygen metabolites, total antioxidant capacity, haptoglobin, oxygen consumption, body mass and cloacal temperature. Lysozyme supplementation intensified the lipopolysaccharide-induced inflammatory response and generated short-term oxidative and metabolic costs. We identified significant interactions between immune supplementation and immune challenge in terms of reactive oxygen metabolites, haptoglobin and oxygen consumption. Our study provides alternative interpretations of differences in oxidative and immunological indices and demonstrates that these indices can also fluctuate and interact across very short time scales, reflecting something akin to current 'health status' or 'physiological condition'. These ephemeral effects highlight the need to broadly consider current physiological condition when drawing conclusions that relate physiology to ecology and evolution.
Product Description
Roche
Bovine Serum Albumin, blood typing: suitable, Endonuclease, Exonuclease, Rnases, Proteinases free