Merck
  • Accelerated detection of viral particles by combining AC electric field effects and micro-Raman spectroscopy.
Accelerated detection of viral particles by combining AC electric field effects and micro-Raman spectroscopy.
Sensors (Basel, Switzerland) (2015-01-13)
Matthew Robert Tomkins, David Shiqi Liao, Aristides Docoslis
ABSTRACT
A detection method that combines electric field-assisted virus capture on antibody-decorated surfaces with the "fingerprinting" capabilities of micro-Raman spectroscopy is demonstrated for the case of M13 virus in water. The proof-of-principle surface mapping of model bioparticles (protein coated polystyrene spheres) captured by an AC electric field between planar microelectrodes is presented with a methodology for analyzing the resulting spectra by comparing relative peak intensities. The same principle is applied to dielectrophoretically captured M13 phage particles whose presence is indirectly confirmed with micro-Raman spectroscopy using NeutrAvidin-Cy3 as a labeling molecule. It is concluded that the combination of electrokinetically driven virus sampling and micro-Raman based signal transduction provides a promising approach for time-efficient and in situ detection of viruses.