Merck
  • Stability of HIB-Cul3 E3 ligase adaptor HIB Is Regulated by Self-degradation and Availability of Its Substrates.
Stability of HIB-Cul3 E3 ligase adaptor HIB Is Regulated by Self-degradation and Availability of Its Substrates.
Scientific reports (2015-08-13)
Zizhang Zhou, Congyu Xu, Ping Chen, Chen Liu, Shu Pang, Xia Yao, Qing Zhang
ABSTRACT
The HIB-Cul3 complex E3 ligase regulates physiological homeostasis through regulating its substrate stability and its activity can be modulated by changing HIB abundance. However, regulation of HIB remains elusive. Here we provide evidence that HIB is degraded through the proteasome by Cul3-mediated polyubiquitination in K48 manner in Drosophila. Strikingly, HIB is targeted for degradation by itself. We further identify that three degrons ((52)LKSS(56)T, (76)LDEE(80)S and (117)MESQ(121)R) and K185 and K198 of HIB are essential for its auto-degradation. Finally, we demonstrate that HIB-Cul3 substrates, Ci and Puc, can effectively protect HIB from HIB-Cul3-mediated degradation. Taken together, our study indicates that there is an exquisite equilibrium between the adaptor and targets to achieve the tight control of the HIB, which is essential for maintaining suitable Hh and JNK signaling. And the mechanism of adaptor self-degradation and reciprocal control of the abundance between adaptor and its substrates is also applied to BTB-Cul3 E3 ligase adaptor dKeap1, dDiablo and dKLHL18.