Merck
Synthesis of potent oxindole CDK2 inhibitors.
Bioorganic & medicinal chemistry (2003-03-28)
Apos Dermatakis, Kin Chun Luk, Wanda DePinto
ABSTRACT
A series of oxindole CDK2 inhibitors was synthesized. These novel analogues have a saturated monosubstituted cyclic moiety at their C-4 position that mimics the ribofuranoside of ATP. This substitution afforded agents with increased potency relative to the parent indolinone and nanomolar range IC(50) against the CDK2 enzyme and two cancer cell lines.
Sigma-Aldrich
4-(N-Boc-amino)piperidine, 96%