Merck
  • 1-[[(3-hydroxy-1-adamantyl)amino]acetyl]-2-cyano-(S)-pyrrolidine: a potent, selective, and orally bioavailable dipeptidyl peptidase IV inhibitor with antihyperglycemic properties.
1-[[(3-hydroxy-1-adamantyl)amino]acetyl]-2-cyano-(S)-pyrrolidine: a potent, selective, and orally bioavailable dipeptidyl peptidase IV inhibitor with antihyperglycemic properties.
Journal of medicinal chemistry (2003-06-13)
Edwin B Villhauer, John A Brinkman, Goli B Naderi, Bryan F Burkey, Beth E Dunning, Kapa Prasad, Bonnie L Mangold, Mary E Russell, Thomas E Hughes
ABSTRACT
Dipeptidyl peptidase IV (DPP-IV) inhibition has the potential to become a valuable therapy for type 2 diabetes. The synthesis and structure-activity relationship of a new DPP-IV inhibitor class, N-substituted-glycyl-2-cyanopyrrolidines, are described as well as the path that led from clinical development compound 1-[2-[5-cyanopyridin-2-yl)amino]ethylamino]acetyl-2-cyano-(S)-pyrrolidine (NVP-DPP728, 8c) to its follow-up, 1-[[(3-hydroxy-1-adamantyl) amino]acetyl]-2-cyano-(S)-pyrrolidine (NVP-LAF237, 12j). The pharmacological profile of 12j in obese Zucker fa/fa rats along with pharmacokinetic profile comparison of 8c and 12j in normal cynomolgus monkeys is discussed. The results suggest that 12j is a potent, stable, selective DPP-IV inhibitor possessing excellent oral bioavailability and potent antihyperglycemic activity with potential for once-a-day administration.
Sigma-Aldrich
3-Amino-1-adamantanol, 96%