- Chemical reduction and spectrophotometric determination of silver, copper and nickel.
Chemical reduction and spectrophotometric determination of silver, copper and nickel.
Talanta (1987-05-01)
G Al-Jabari, B Jaselskis
PMID18964340
ABSTRACT
Silver(I), copper(II) and nickel(II) can be reduced to the metallic state by formaldehyde at pH 11, chromium(II) in 2.5M sulphuric acid, and borohydride at pH 5.5-6.0, respectively. Reoxidation of these metals with iron(III) in the presence of Ferrozine enables their determination at concentration below 1 mug/ml by measurement of the absorbance of the iron(II)-Ferrozine complex at 562 nm, with a precision better than 3%. The apparent molar absorptivities for silver, copper and nickel are 2.78 x 10(4), 5.56 x 10(4) and 5.58 x 10(4) l.mole(-1).cm(-1), respectively. The average thickness of silver films on glass surfaces can be determined in the way.
Sigma-Aldrich
3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-p,p′-disulfonic acid monosodium salt hydrate, 97%