Merck
  • Electronic spectra of 2- and 3-tolunitrile in the gas phase. I. A study of methyl group internal rotation via rovibronically resolved spectroscopy.
Electronic spectra of 2- and 3-tolunitrile in the gas phase. I. A study of methyl group internal rotation via rovibronically resolved spectroscopy.
The Journal of chemical physics (2016-02-02)
José Arturo Ruiz-Santoyo, Josefin Wilke, Martin Wilke, John T Yi, David W Pratt, Michael Schmitt, Leonardo Álvarez-Valtierra
ABSTRACT
Rotationally resolved fluorescence excitation spectra of the origin bands in the S1 ← S0 transition of 2-tolunitrile (2TN) and 3-tolunitrile (3TN) have been recorded in the collision-free environment of a molecular beam. Analyses of these data provide the rotational constants of each molecule and the potential energy curves governing the internal rotation of the attached methyl groups in both electronic states. 2TN exhibits much larger barriers along this coordinate than 3TN. Interestingly, the electronic transition dipole moment in both molecules is markedly influenced by the position of the attached methyl group rather than the position of the cyano group; possible reasons for this intriguing behavior are discussed.
Sigma-Aldrich
o-Tolunitrile, ≥97.0% (GC)