Merck
  • C-reactive protein, interleukin 6, and risk of developing type 2 diabetes mellitus.
C-reactive protein, interleukin 6, and risk of developing type 2 diabetes mellitus.
JAMA (2001-07-24)
A D Pradhan, J E Manson, N Rifai, J E Buring, P M Ridker
ABSTRACT
Inflammation is hypothesized to play a role in development of type 2 diabetes mellitus (DM); however, clinical data addressing this issue are limited. To determine whether elevated levels of the inflammatory markers interleukin 6 (IL-6) and C-reactive protein (CRP) are associated with development of type 2 DM in healthy middle-aged women. Prospective, nested case-control study. The Women's Health Study, an ongoing US primary prevention, randomized clinical trial initiated in 1992. From a nationwide cohort of 27 628 women free of diagnosed DM, cardiovascular disease, and cancer at baseline, 188 women who developed diagnosed DM over a 4-year follow-up period were defined as cases and matched by age and fasting status with 362 disease-free controls. Incidence of confirmed clinically diagnosed type 2 DM by baseline levels of IL-6 and CRP. Baseline levels of IL-6 (P<.001) and CRP (P<.001) were significantly higher among cases than among controls. The relative risks of future DM for women in the highest vs lowest quartile of these inflammatory markers were 7.5 for IL-6 (95% confidence interval [CI], 3.7-15.4) and 15.7 for CRP (95% CI, 6.5-37.9). Positive associations persisted after adjustment for body mass index, family history of diabetes, smoking, exercise, use of alcohol, and hormone replacement therapy; multivariate relative risks for the highest vs lowest quartiles were 2.3 for IL-6 (95% CI, 0.9-5.6; P for trend =.07) and 4.2 for CRP (95% CI, 1.5-12.0; P for trend =.001). Similar results were observed in analyses limited to women with a baseline hemoglobin A(1c) of 6.0% or less and after adjustment for fasting insulin level. Elevated levels of CRP and IL-6 predict the development of type 2 DM. These data support a possible role for inflammation in diabetogenesis.