Merck
  • Association between polymorphism of the interleukin-13 gene and susceptibility to hepatocellular carcinoma in the Chinese population.
Association between polymorphism of the interleukin-13 gene and susceptibility to hepatocellular carcinoma in the Chinese population.
PloS one (2015-02-07)
Yan Deng, Ming Xie, Li Xie, Jian Wang, Taijie Li, Yu He, Ruolin Li, Shan Li, Xue Qin
ABSTRACT
Interleukin-13 (IL-13) is a potent pleiotropic cytokine that is produced by activated CD4 T cells. This study was undertaken to determine the relationship between two IL-13 gene single nucleotide polymorphisms (SNP rs1800925 and SNP rs20541) and the incidence of hepatitis B virus-related (HBV) hepatocellular carcinoma (HCC). Three hundred and ninety-eight HBV-positive individuals (192 HCC and 206 patients with chronic hepatitis) and one hundred and ninety-two healthy participants from the First Affiliated Hospital of Guangxi Medical University were enrolled in this study. The results showed no significant differences between the genotype and allele frequencies of the IL-13 gene rs1800925 and rs20541 polymorphisms and chronic hepatitis B risk after adjusting for age, sex, tobacco use, and alcohol intake using binary logistic regression analyses. Regarding the rs20541 SNP, the GA genotype was significantly related to a decreased risk of HCC after adjusting for age, sex, tobacco use, and alcohol intake using binary logistic regression analyses (The odds ratio (OR) = 0.54, 95% confidence intervals (CI) 0.34-0.87). The adjusted OR for the GA and AA genotypes combined was 0.68 (95% CI 0.39-0.90). This study indicates that the functional IL-13 rs20541 polymorphism may contribute to the risk of HCC and that the rs20541 polymorphism is a protective factor for HCC.
Sigma-Aldrich
IL-13 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)