Merck
  • IL-33 Signaling Protects from Murine Oxazolone Colitis by Supporting Intestinal Epithelial Function.
IL-33 Signaling Protects from Murine Oxazolone Colitis by Supporting Intestinal Epithelial Function.
Inflammatory bowel diseases (2015-08-28)
Amanda Waddell, Jefferson E Vallance, Preston D Moore, Amy T Hummel, David Wu, Shiva K Shanmukhappa, Lin Fei, M Kay Washington, Phillip Minar, Lori A Coburn, Susumu Nakae, Keith T Wilson, Lee A Denson, Simon P Hogan, Michael J Rosen
ABSTRACT
IL-33, a member of the IL-1 cytokine family that signals through ST2, is upregulated in ulcerative colitis (UC); however, the role of IL-33 in colitis remains unclear. IL-33 augments type 2 immune responses, which have been implicated in UC pathogenesis. We sought to determine the role of IL-33 signaling in oxazolone (OXA) colitis, a type 2 cytokine-mediated murine model of UC. Colon mucosal IL-33 expression was compared between pediatric and adult UC and non-IBD patients using immunohistochemistry and real-time PCR. OXA colitis was induced in WT, IL-33, and ST2 mice, and histopathology, cytokine levels, and goblet cells were assessed. Transepithelial resistance was measured across IL-33-treated T84 cell monolayers. Colon mucosal IL-33 was increased in pediatric patients with active UC and in OXA colitis. IL-33 and ST2 OXA mice exhibited increased disease severity compared with WT OXA mice. OXA induced a mixed mucosal cytokine response, but few differences were observed between OXA WT and IL-33 or ST2 mice. Goblet cells were significantly decreased in IL-33 and ST2 OXA compared with WT OXA mice. IL-33 augmented transepithelial resistance in T84 cells, and this effect was blocked by the ERK1/2 inhibitor PD98,059. OXA colitis is exacerbated in IL-33 and ST2 mice. Increased mucosal IL-33 in human UC and murine colitis may be a homeostatic response to limit inflammation, potentially through effects on epithelial barrier function. Further investigation of IL-33 protective mechanisms would inform the development of novel therapeutic approaches.
Sigma-Aldrich
4-Ethoxymethylene-2-phenyl-2-oxazolin-5-one, purified by recrystallization